Sodium sulfate decahydrate, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra_pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
1.06642.2500 1.06642.9025
1.06642.2500EA 8750 SEK
1.06642.2500 1.06642.9025
Sodium sulfate decahydrate, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, extra_pure suitable for use as excipient, SAFC®
Natriumsulfat Dekahydrat
Formel: Na₂SO₄·10H₂O
MW: 322.2 g/mol
Kokpunkt: 1429
Smältpunkt: 32.4
Densitet: 1.464 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149189
CAS nummer: 7727-73-3
EINECS: 231-820-9
Merck Index: 13,08755