Natriumtiosulfat pentahydrat ≥99%
Leverantör: Thermo Fisher Scientific
A17914.0C A17914.0E A17914.36
A17914.0CEA 5300 SEK
A17914.0C A17914.0E A17914.36
Natriumtiosulfat pentahydrat ≥99%
Natriumtiosulfat pentahydrat
Purity: 99+% CAS No.: 10102-17-7 Molecular Formula: Na2O3S2.5H2O Formula Weight: 248.17 (158.10anhy) Odour: Odourless pH: 6.5-8.0 Melting Point: 48° Boiling Point: 100° -5H2O Merck Reference: 14,8694 MDL No.: MFCD00149186
Formel: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Kokpunkt: 100 °C (–5H₂O)
Smältpunkt: 48
Densitet: 1.69
Storage Temperature: Omgivande
MDL Number: MFCD00149186
CAS nummer: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844