Natriumdihydrogenorthofosfat dihydrat, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Leverantör: Merck
Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate
M-Clarity™ quality level = MQ300
1.06342.0250 1.06342.1000 1.06342.2500
1.06342.0250EA 462 SEK
1.06342.0250 1.06342.1000 1.06342.2500
Natriumdihydrogenorthofosfat dihydrat, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Natriumdihydrogenorthofosfat dihydrat
Formel: NaH₂PO₄·2H₂O
MW: 156.01 g/mol
Kokpunkt: ∼650 °C (1013 hPa)
Smältpunkt: 60
Densitet: 1.91 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149209
CAS nummer: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806
Order Now
Specifikation testresultat
Solubility 850 g/l (20 °C)
Melting Point 60 °C
Molar Mass 156.02 g/mol
Bulk Density 1000 kg/m3
Density 1.915 g/cm3 (20 °C)
pH 4.5 (12 g/l, H2O, 25 °C)