Natriumdihydrogenorthofosfat monohydrat, EMPROVE® EXPERT USP, BP, extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
1.06349.1000 1.06349.9025 1.06349.9014
1.06349.1000EA 2675 SEK
1.06349.1000 1.06349.9025 1.06349.9014
Natriumdihydrogenorthofosfat monohydrat, EMPROVE® EXPERT USP, BP, extra pure suitable for use as excipient, SAFC®
Natriumdihydrogenorthofosfat monohydrat
Formel: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Smältpunkt: 100
Densitet: 2.04 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149208
CAS nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM