di-Natriumhydrogen fosfat dodekahydrat, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
1.06573.5000 1.06573.9026 1.06573.9029 1.06573.9500
1.06573.5000EA 6675 SEK
1.06573.5000 1.06573.9026 1.06573.9029 1.06573.9500
di-Natriumhydrogen fosfat dodekahydrat, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
di-Natriumhydrogen fosfat dodekahydrat
Formel: Na₂HPO₄·12H₂O
MW: 358,14 g/mol
Smältpunkt: 35 °C
Densitet: 1,52 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00149181
CAS nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733