tetranatriumdifosfat Dekahydrat 97
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
A16195.0B A16195.0I A16195.30
A16195.0BEA 1900 SEK
A16195.0B A16195.0I A16195.30
tetranatriumdifosfat Dekahydrat 97
tetranatriumdifosfat Dekahydrat
Purity: 97+% CAS No.: 13472-36-1 Molecular Formula: Na4O7P2.10H2O Formula Weight: 446.06 (265.90anhy) Odour: Odourless pH: 9.5-10.5 Melting Point: 79.5° Merck Reference: 14,9240 MDL No.: MFCD00149200
Formel: Na₄P₂O₇·10H₂O
MW: 446.06 g/mol
Smältpunkt: 79.5 °C
Densitet: 1.82
Storage Temperature: Omgivande
MDL Number: MFCD00149200
CAS nummer: 13472-36-1
EINECS: 231-767-1