dinatriumtetraborat Dekahydrat ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
A16176.0C A16176.0E A16176.36
A16176.0CEA 6275 SEK
A16176.0C A16176.0E A16176.36
dinatriumtetraborat Dekahydrat ≥99%
dinatriumtetraborat Dekahydrat
Purity: 99+% CAS No.: 1303-96-4 Molecular Formula: B4Na2O7.10H2O Formula Weight: 381.37 (201.22anhy) Odour: Odourless pH: 9,24 Melting Point: 75° Merck Reference: 14,8590 MDL No.: MFCD00149193
Formel: Na₂B₄O₇·10H₂O
MW: 381,37 g/mol
Kokpunkt: 320 °C (1013 hPa)
Smältpunkt: 75 °C
Densitet: 1,73
Storage Temperature: Omgivande
MDL Number: MFCD00149193
CAS nummer: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 14,8590