Blydiacetat trihydrat ≥99.995%, Puratronic®
Leverantör: Thermo Scientific
Fara
Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
10719.06 10719.18 10719.30
10719.06EA 540 SEK
10719.06 10719.18 10719.30
Blydiacetat trihydrat ≥99.995%, Puratronic®
Blydiacetat trihydrat
Purity: 99.995% CAS No.: 6080-56-4 Molecular Formula: Pb(OOCCH3)2.3H2O Formula Weight: 379.33 (325.28anhy) Physical Form: Crystalline Density: 2.55 Melting Point: 75° Beilstein Reference: 3730298 Merck Reference: 14,5397 UN No.: 1616 MDL No.: MFCD00150023
Formel: (H₃CCOO)₂Pb·3H₂O
MW: 379,34 g/mol
Kokpunkt: 280 °C (1013 hPa)
Smältpunkt: 75 °C
Densitet: 2,55
Storage Temperature: Omgivande
MDL Number: MFCD00150023
CAS nummer: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 14,5397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=cb30c0c27f7a30685c4396352ba26dc492c2aba5&prid=web&clid=SAPDAM