Blydiacetat trihydrat 99.999%, extra pure
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
317230250 317230050
ACRO317230250EA 2975 SEK
ACRO317230250 ACRO317230050
Blydiacetat trihydrat 99.999%, extra pure
Blydiacetat trihydrat
Formel: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Kokpunkt: 280
Smältpunkt: 75
Storage Temperature: Omgivande
MDL Number: MFCD00150023
CAS nummer: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals
Total trace metal impurities ≤0.001 %