Natriumtiosulfat 0.1 mol/l (0.1 N) aqueous solution, AVS TITRINORM Volumetric
Leverantör: VWR Chemicals
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
31553.294EA 165 SEK
31553.294 31553.363 31553.408
Natriumtiosulfat 0.1 mol/l (0.1 N) aqueous solution, AVS TITRINORM Volumetric
Natriumtiosulfat
Formel: Na₂S₂O₃
MW: 158.11 g/mol
Kokpunkt: 100
Smältpunkt: 48
Storage Temperature: Omgivande
MDL Number: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844
Order Now
Specifikation testresultat
Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed