Natriumtiosulfat 0.1 mol/l (0.1 N) aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverantör: Avantor
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
7101.1000EA 1075 SEK
7101.1000 7101.9010 7101.9020
Natriumtiosulfat 0.1 mol/l (0.1 N) aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Natriumtiosulfat
Formel: Na₂S₂O₃
MW: 158.11 g/mol
Kokpunkt: 100
Smältpunkt: 48
Storage Temperature: Omgivande
MDL Number: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844
Order Now
Specifikation testresultat
Titer (mol/l) 0.0997 - 0.1003
Trace Impurities (in ppm)
Heavy Metals (as Pb) max. 1
Sulfate and Sulfite (as SO4) max. 50