Natriumhydroxid 0,1 mol/l (0,1 N) i vattenlösning, Titripur® Reag. Ph. Eur. volumetrisk lösning, Supelco®
Leverantör: Merck
Synonyms: Natronlut, Kaustik soda
M-Clarity™ quality level = MQ100
1.09141.1000 1.09141.5000 1.09141.9010 1.09141.9025 1.09141.4000 1.09141.1003 1.09141.9013
1.09141.1000EA 211.96 SEK
1.09141.1000 1.09141.5000 1.09141.9010 1.09141.9025 1.09141.4000 1.09141.1003 1.09141.9013
Natriumhydroxid 0,1 mol/l (0,1 N) i vattenlösning, Titripur® Reag. Ph. Eur. volumetrisk lösning, Supelco®
Natriumhydroxid
Merck have introduced into their Titripur® Titrants range packs with additional technology - Supelco® SmartTitrants use an RFID tag to provide seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. Mettler Toledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.
Formel: NaOH
MW: 40 g/mol
Kokpunkt: 1390
Smältpunkt: 323
Densitet: 1.00 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now
Specifikation testresultat
Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.