Salpetersyra standardised solution 1.0 N
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Nitric acid solution
35624.AP 35624.K2
35624.APEA 950 SEK
35624.AP 35624.K2
Salpetersyra standardised solution 1.0 N
Salpetersyra
CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colourless UN No.: 3264 MDL No.: MFCD00011349
Formel: HNO₃
MW: 63.01 g/mol
Kokpunkt: 87
Smältpunkt: -42
Densitet: 1.035 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610