Saltsyra 0.2 mol/l (0.2 N), AVS TITRINORM Volumetric
Leverantör: VWR Chemicals
Varning
Synonyms: Hydrochloric acid solution, Muriatic acid
31983.290EA 204 SEK
31983.290 31983.370
Saltsyra 0.2 mol/l (0.2 N), AVS TITRINORM Volumetric
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Densitet: 1.004 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III
Order Now
Specifikation testresultat
Titer (20°C; real value 0.2 % accuracy) 0.1996 - 0.2004 mol/l
NIST traceable Confirmed