Saltsyra 0.1 mol/l (0.1 N) Volumetric
Leverantör: REAGECON
Synonyms: Hydrochloric acid solution, Muriatic acid
H20105 H20101
REGEH20105EA 870 SEK
REGEH20105 REGEH20101
Saltsyra 0.1 mol/l (0.1 N) Volumetric
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Densitet: 1.004 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III