(±)-1-Aminoindan 98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
1-Aminoindan, 1-Indanamine, 2,3-Dihydro-1H-inden-1-amine, (±)-1-Indanylamine
103850010
103850050
ACRO103850010EA
1150
SEK
ACRO103850010
ACRO103850050
(±)-1-Aminoindan 98%
(±)-1-Aminoindan
Formel:
C₉H₁₁N MW: 133.19 g/mol Kokpunkt: 96…97 °C (8 mmHg) Smältpunkt: 1,5 Densitet: 1.038 g/cm³ (20 °C) Flampunkt: 94 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00003799 CAS nummer: 34698-41-4 EINECS: 252-158-7 |
Specifikation testresultat
Appearance | Clear colorless to light yellow to green Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5600 to 1.5640 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...