DL-kamfer 96%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone
164640010 164640051 164642500
ACRO164640010EA 1375 SEK
ACRO164640010 ACRO164640051 ACRO164642500
DL-kamfer 96%
DL-kamfer
Formel: C₁₀H₁₆O
Kokpunkt: 204 °C (760 mmHg)
Smältpunkt: 175…177 °C
Densitet: 0,992 g/cm³ (25 °C)
Flampunkt: 64 °C
Storage Temperature: Omgivande
MDL Number: MFCD00074738
CAS nummer: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III
Order Now
Specifikation testresultat
Appearance White Crystals and/or chunks
Infrared spectrum Conforms
GC ≥95 %
Residue after evaporation ≤0.1 %
Angle of optical rotation -0.15° to +0.15° (20°C, 589 nm) (c=10, alcohol)