1,2-Propanediol, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
1.07478.2500 1.07478.6025
1.07478.2500EA 7000 SEK
1.07478.2500 1.07478.6025
1,2-Propanediol, EMPROVE® EXPERT BP, Ph. Eur., USP suitable for use as excipient, SAFC®
(±) -1,2-propandiol
Formel: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Kokpunkt: 187.6
Smältpunkt: –59 °C
Densitet: 1.04 g/cm³ (20 °C)
Flampunkt: 99
Storage Temperature: Omgivande
MDL Number: MFCD00064272
CAS nummer: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM