(±)-3-Octanol 97%
Leverantör: Thermo Fisher Scientific
Varning
203770500 203772500
ACRO203770500EA 1175 SEK
ACRO203770500 ACRO203772500
(±)-3-Octanol 97%
(±)-3-Octanol
Formel: C₈H₁₈O
Kokpunkt: 174…176 °C (760 mmHg)
Smältpunkt: –45 °C
Densitet: 0,825 g/cm³
Flampunkt: 67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004590
CAS nummer: 589-98-0
EINECS: 209-667-4
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4320 (20°C, 589 nm)
Density (20°C) 0.817 to 0.827 kg/l
GC ≥96.0 %