(±)-2-Pentanol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(±)-Methyl propyl carbinol, (±)-2-Hydroxy pentane, DL-2-Pentanol, (±)-Propyl methyl carbinol
P0056-25ML
P0056-500ML
TCIAP0056-25MLEA
255
SEK
TCIAP0056-25ML
TCIAP0056-500ML
(±)-2-Pentanol ≥98.0%
(±)-2-Pentanol
Formel:
C₅H₁₂O MW: 88,15 g/mol Kokpunkt: 118°C Smältpunkt: –50 °C Densitet: 0,81 g/cm³ (20 °C) Flampunkt: 34°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004579 CAS nummer: 6032-29-7 EINECS: 227-907-6 UN: 1105 ADR: 3,III Merck Index: 14,07119 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...