Fosforsyra 85% ≥85%, EMPROVE® ESSENTIAL JPE, BP, E338, Ph. Eur., NF, extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: Phosphoric acid
1.00563.1000 1.00563.2500 1.00563.9025 1.00563.9950 1.00563.9180 1.00563.5000
1.00563.1000EA 4600 SEK
1.00563.1000 1.00563.2500 1.00563.9025 1.00563.9950 1.00563.9180 1.00563.5000
Fosforsyra 85% ≥85%, EMPROVE® ESSENTIAL JPE, BP, E338, Ph. Eur., NF, extra pure suitable for use as excipient, SAFC®
Fosforsyra
Formel: H₃PO₄
MW: 98 g/mol
Kokpunkt: 158
Smältpunkt: 21
Densitet: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011340
CAS nummer: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344