Saltsyra 25%, EMPROVE® ESSENTIAL Ph. Helv., extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: Hydrochloric acid solution, Muriatic acid
1.00312.2500 1.00312.9025 1.00312.9180
1.00312.2500EA 1925 SEK
1.00312.2500 1.00312.9025 1.00312.9180
Saltsyra 25%, EMPROVE® ESSENTIAL Ph. Helv., extra pure suitable for use as excipient, SAFC®
Saltsyra
Formel: HCl
Kokpunkt: 110 °C (1013 hPa)
Smältpunkt: –30 °C
Densitet: 1,18 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II