Pyridin-D5 99 atom % D for NMR spectroscopy
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Pyridine-[²H₅]
166570050 166570100 166570250
ACRO166570050EA 1550 SEK
ACRO166570050 ACRO166570100 ACRO166570250
Pyridin-D5 99 atom % D for NMR spectroscopy
Pyridin-D5
Formel: C₅D₅N
Kokpunkt: 114,4 °C (1013 hPa)
Smältpunkt: –41 °C
Densitet: 1,05 g/cm³ (20 °C)
Flampunkt: 20 °C
Storage Temperature: Omgivande
MDL Number: MFCD00044639
CAS nummer: 7291-22-7
EINECS: 230-720-2
UN: 1282
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤10 %
D-Enrichment ≥98.5 %
Specific optical rotation +7° to +8° (20°C, 589 nm) c=5 in water