Tetrahydrofuran-D8 99.5 atom % D for NMR spectroscopy
Leverantör: Thermo Scientific
Fara
Synonyms: Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8
180130010 180130050 180130100
ACRO180130010EA 1275 SEK
ACRO180130010 ACRO180130050 ACRO180130100
Tetrahydrofuran-D8 99.5 atom % D for NMR spectroscopy
Tetrahydrofuran-D8
Formel: C₄D₈O
MW: 80,04 g/mol
Kokpunkt: 65…66 °C (760 mmHg)
Smältpunkt: –106 °C
Densitet: 0,98 g/cm³
Flampunkt: –21 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00044238
CAS nummer: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.06 % (Coulometric)
D-Enrichment ≥99.4 %
Titration with HClO4 ≥98.5 % (on dry substance)