n-Hexan-D14 99+ atom % D for NMR spectroscopy
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Hexane-D14, n-Hexane-[²H₁₄]
244010010 244010050
ACRO244010010EA 2750 SEK
ACRO244010010 ACRO244010050
n-Hexan-D14 99+ atom % D for NMR spectroscopy
n-Hexan-D14
Formel: D₃C(CD₂)₄CD₃
Kokpunkt: 69 °C (1013 hPa)
Densitet: 0,96 g/cm³ (20 °C)
Flampunkt: –22 °C
Storage Temperature: Omgivande
MDL Number: MFCD00037562
CAS nummer: 21666-38-6
UN: 1208
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
D-Enrichment ≥99.0 %