Metanol-D3 99.5 atom % D for NMR spectroscopy
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Carbinol-D3, Hydroxy methane-D3, Methanol-[²H₃], Wood alcohol-D3, Methyl alcohol-D3
300730025 300730100
ACRO300730025EA 1675 SEK
ACRO300730025 ACRO300730100
Metanol-D3 99.5 atom % D for NMR spectroscopy
Metanol-D3
Formel: D₃COH
MW: 35.02 g/mol
Kokpunkt: 65 °C (760 mmHg)
Smältpunkt: -98
Densitet: 0.867 g/cm³
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00055877
CAS nummer: 1849-29-2
EINECS: 217-435-9
UN: 1230
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥99.4 %