Kloroform-D1 (99.8% D) for NMR spectroscopy, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
M-Clarity™ quality level = MQ200
1.02450.0025 1.02450.0100 1.02450.0500
1.02450.0025EA 426 SEK
1.02450.0025 1.02450.0100 1.02450.0500
Kloroform-D1 (99.8% D) for NMR spectroscopy, Sigma-Aldrich®
Kloroform-D1
Formel: CDCl₃
Kokpunkt: 60,8 °C (1013 hPa)
Densitet: 1,5 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00000827
CAS nummer: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III