Kloroform-D1 100.0 atom % D for NMR spectroscopy
Leverantör: Thermo Scientific
Fara
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
166270100 166270500 166271000
ACRO166270100EA 1075 SEK
ACRO166270100 ACRO166270500 ACRO166271000
Kloroform-D1 100.0 atom % D for NMR spectroscopy
Kloroform-D1
Formel: CDCl₃
MW: 120,38 g/mol
Kokpunkt: 60,8 °C (1013 hPa)
Smältpunkt: –64 °C
Densitet: 1,5 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00000827
CAS nummer: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.01 % (Coulometric)
D-Enrichment ≥99.95 %
GC ≥96.0 %