Diklormetan-D2 99.8 atom % D for NMR spectroscopy
Leverantör: Thermo Scientific
Varning
Synonyms: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2
321270050 321270100
ACRO321270050EA 2050 SEK
ACRO321270050 ACRO321270100
Diklormetan-D2 99.8 atom % D for NMR spectroscopy
Diklormetan-D2
Formel: CD₂Cl₂
MW: 86,92 g/mol
Kokpunkt: 40 °C (1013 hPa)
Smältpunkt: –97 °C
Densitet: 1,36 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000882
CAS nummer: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %