Aceton-D6 99+ atom % D for NMR spectroscopy
Leverantör: Thermo Fisher Scientific
Fara
174900100 174900500
ACRO174900100EA 965 SEK
ACRO174900100 ACRO174900500
Aceton-D6 99+ atom % D for NMR spectroscopy
Aceton-D6
Formel: D₃CC(O)CD₃
Kokpunkt: 55,5 °C (1013 hPa)
Smältpunkt: –93 °C
Densitet: 0,87 g/cm³
Flampunkt: –20 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00044635
CAS nummer: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥99.0 %
HPLC ≥97.5 %