1-Bromo-4-methylcyclohexane cis- and trans-mixture ≥97%
Leverantör: Thermo Fisher Scientific
L02640.06 L02640.14
L02640.06EA 1650 SEK
L02640.06 L02640.14
1-Bromo-4-methylcyclohexane cis- and trans-mixture ≥97%
1-Bromo-4-methylcyclohexane
Purity: 97% CAS No.: 6294-40-2 Molecular Formula: C7H13Br Formula Weight: 177.09 Refractive Index: 1.4880 Boiling Point: 94-96°/50mm MDL No.: MFCD00014282
Formel: C₇H₁₃Br
MW: 177.08 g/mol
Kokpunkt: 94…96 °C (50 mmHg)
Densitet: 1.262
Storage Temperature: Omgivande
MDL Number: MFCD00014282
CAS nummer: 6294-40-2
EINECS: 228-559-8