2-Octene cis- and trans-mixture ≥98%
Leverantör: Thermo Scientific
Fara
L03862.06 L03862.14
L03862.06EA 805 SEK
L03862.06 L03862.14
2-Octene cis- and trans-mixture ≥98%
2-Octene
Purity: 98% CAS No.: 111-67-1 Molecular Formula: C8H16 Formula Weight: 112.22 Flash Point: 14°(57°F) Refractive Index: 1.4150 Boiling Point: 123-125° Beilstein Reference: 1719495 UN No.: 3295 MDL No.: MFCD00009532
Formel: C₈H₁₆
MW: 112,22 g/mol
Kokpunkt: 123…125 °C
Smältpunkt: –106,8 °C
Densitet: 0,715
Flampunkt: 14 °C (57 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009532
CAS nummer: 111-67-1
EINECS: 203-894-2
UN: 3295
ADR: 3,II