2-Bromo-2-butene cis- and trans-mixture ≥98% stabilised
Leverantör: Thermo Scientific
Fara
A18058.14 A18058.22
A18058.14EA 1600 SEK
A18058.14 A18058.22
2-Bromo-2-butene cis- and trans-mixture ≥98% stabilised
2-Bromo-2-butene
Stabilized
Purity: 98% CAS No.: 13294-71-8 Molecular Formula: C4H7Br Formula Weight: 135.00 Storage Temperature: 2°C to 8°C Flash Point: 1°(33°F) Refractive Index: 1.4610 Odour: Recognizable Boiling Point: 82-90° Beilstein Reference: 1733329 UN No.: 1993 MDL No.: MFCD00000141
Formel: C₄H₇Br
MW: 135 g/mol
Kokpunkt: 82…90 °C
Densitet: 1,328
Flampunkt: 1 °C (33 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00000141
CAS nummer: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3,II