Natriumoktan-1-sulfonat, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: Sodium octane-1-sulphonate
M-Clarity™ quality level = MQ200
1.12292.0005 1.12292.0025 1.12292.0002
1.12292.0005EA 2230 SEK
1.12292.0005 1.12292.0025 1.12292.0002
Natriumoktan-1-sulfonat, Sigma-Aldrich®
Natriumoktan-1-sulfonat
Formel: C₈H₁₇NaO₃S
MW: 216,28 g/mol
Smältpunkt: > 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007544
CAS nummer: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=9b17758a2a1f520c7918ded6d9f3eba0597d62e9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 216.27 g/mol
Bulk Density 345 kg/m3
pH 5.5 - 7.5 (100 g/l, H2O, 20 °C)