Krom ≥99.995%, pieces, Thickness: 2-3 mm (0.08-0.12 in)
Leverantör: Thermo Scientific
38494.18 38494.30
38494.18EA 2850 SEK
38494.18 38494.30
Krom ≥99.995%, pieces, Thickness: 2-3 mm (0.08-0.12 in)
Krom
Purity: 99.995% CAS No.: 7440-47-3 Physical Form: Broken plate, Large irregular pieces Odour: Odourless MDL No.: MFCD00010944
Formel: Cr
MW: 52 g/mol
Kokpunkt: 2469 °C (1013 hPa)
Smältpunkt: 1840 °C
Densitet: 7,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010944
CAS nummer: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288