Krom 99,997 Metaller basis, pieces, Puratronic® ≤ 6 mm Electrolytic
Leverantör: Thermo Fisher Scientific
10151.14 10151.22
10151.14EA 4200 SEK
10151.14 10151.22
Krom 99,997 Metaller basis, pieces, Puratronic® ≤ 6 mm Electrolytic
Krom
Purity: 99.997% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944
Formel: Cr
MW: 52 g/mol
Kokpunkt: 2469
Smältpunkt: 1840
Densitet: 7.2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010944
CAS nummer: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288