Krom ≥99.97%, kuber, Length: 12.5 mm (0.492 in)
Leverantör: Thermo Scientific
10149.KT 10149.KU
10149.KTEA 5475 SEK
10149.KT 10149.KU
Krom ≥99.97%, kuber, Length: 12.5 mm (0.492 in)
Krom
Purity: 99.97% CAS No.: 7440-47-3 Physical Form: ≈14.7g/pc Hot Isostatically Pressed Odour: Odourless MDL No.: MFCD00010944
Formel: Cr
MW: 52 g/mol
Kokpunkt: 2469 °C (1013 hPa)
Smältpunkt: 1840 °C
Densitet: 7,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010944
CAS nummer: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288