Zink 98+% stable (acc. to UN classification class 4), dust
Leverantör: Thermo Scientific
Varning
198340010 198340025 198341000 198340100 198340250
ACRO198340010EA 925 SEK
ACRO198340010 ACRO198340025 ACRO198341000 ACRO198340100 ACRO198340250
Zink 98+% stable (acc. to UN classification class 4), dust
Zink
Stable acc. to UN classification class 4
Formel: Zn
MW: 65,39 g/mol
Kokpunkt: 906 °C (760 mmHg)
Smältpunkt: 419 °C
Densitet: 7,14 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Grey Powder
Particle Size/Sieve analysis ≤0.1 % Residue on 250 µm(+60 mesh) sieve
Zinc (Zn) ≥98.0 %