Zink ≥99.9985%, folie, Puratronic®, Thickness: 1.0 mm (0.04 in)
Leverantör: Thermo Fisher Scientific
11915.FF 11915.FI 11915.GH
11915.FFEA 2625 SEK
11915.FF 11915.FI 11915.GH
Zink ≥99.9985%, folie, Puratronic®, Thickness: 1.0 mm (0.04 in)
Zink
Purity: 99.9985% CAS No.: 7440-66-6 Physical Form: ≈4.5g/25x25mm Odour: Odourless MDL No.: MFCD00011291
Formel: Zn
Kokpunkt: 908 °C (1013 hPa)
Smältpunkt: 420 °C
Densitet: 7,14 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255