Zink ≥99.98%, folie, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Fisher Scientific
10436.BS 10436.BW 10436.CB
10436.BSEA 1550 SEK
10436.BS 10436.BW 10436.CB
Zink ≥99.98%, folie, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Zink
Purity: 99.98% CAS No.: 7440-66-6 Physical Form: ≈53g/30x10cm Odour: Odourless MDL No.: MFCD00011291
Formel: Zn
MW: 65.39 g/mol
Kokpunkt: 908
Smältpunkt: 420
Densitet: 7.14 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255
Order Now
Specifikation testresultat
Appearance (Color) Gray
Form Foil
Comment 0.22 - 0.28 mm thick
Width 27 - 33 mm
Total Metal Impurities 0.02% max.