Zink, granular 20 mesh
Leverantör: Thermo Fisher Scientific
Fara
222601000 222605000 222600025
ACRO222601000EA 460 SEK
ACRO222601000 ACRO222605000 ACRO222600025
Zink, granular 20 mesh
Zink
Formel: Zn
Kokpunkt: 908 °C (1013 hPa)
Smältpunkt: 420 °C
Densitet: 7,14 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
UN: 1436
Merck Index: 12,10255
Order Now
Specifikation testresultat
Appearance Silver grey Granules or filings
Titration Complexometric ≥99.8 %
Particle size 20 mesh
Suitability (For determination of As) Passes test
Iron (Fe) ≤0.01 %
Lead (Pb) ≤0.01 %