Koppar ≥99.9999%, folie, Puratronic®, Thickness: 0.5 mm (0.02 in)
Leverantör: Thermo Fisher Scientific
42975.FI 42975.GH
42975.FIEA 4150 SEK
42975.FI 42975.GH
Koppar ≥99.9999%, folie, Puratronic®, Thickness: 0.5 mm (0.02 in)
Koppar
Purity: 99.9999% CAS No.: 7440-50-8 Physical Form: ≈11.2g/50x50mm Odour: Odourless MDL No.: MFCD00010965
Formel: Cu
MW: 63.55 g/mol
Kokpunkt: 2595
Smältpunkt: 1083
Densitet: 8.94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545