Koppar 99 Metaller basis, powder -625 mesh (APS 0,50-1,5 µm)
Leverantör: Thermo Fisher Scientific
Varning
Argon_filled
41205.09 41205.18 41205.30
41205.09EA 1550 SEK
41205.09 41205.18 41205.30
Koppar 99 Metaller basis, powder -625 mesh (APS 0,50-1,5 µm)
Koppar
Purity: 99% CAS No.: 7440-50-8 Physical Form: Packaged under argon Appearance: Reddish Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965
Formel: Cu
MW: 63.55 g/mol
Kokpunkt: 2595
Smältpunkt: 1083
Densitet: 8.94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545