Koppar 99 Metaller basis, powder -325 mesh, 10% max +325 mesh
Leverantör: Thermo Fisher Scientific
Varning
13990.18 13990.36 13990.A3 13990.A9
13990.18EA 1125 SEK
13990.18 13990.36 13990.A3 13990.A9
Koppar 99 Metaller basis, powder -325 mesh, 10% max +325 mesh
Koppar
Purity: 99% CAS No.: 7440-50-8 Physical Form: APS 8-11 micron (FSSS) Appearance: Copper Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965
Formel: Cu
MW: 63.55 g/mol
Kokpunkt: 2595
Smältpunkt: 1083
Densitet: 8.94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545