Koppar 99,999 Metaller basis, rods, Puratronic®, Ø 5 mm (0.2 in)
Leverantör: Thermo Fisher Scientific
11444.DO 11444.HT
11444.DOEA 4200 SEK
11444.DO 11444.HT
Koppar 99,999 Metaller basis, rods, Puratronic®, Ø 5 mm (0.2 in)
Koppar
Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈1.75g/cm Odour: Odourless MDL No.: MFCD00010965
Formel: Cu
MW: 63.55 g/mol
Kokpunkt: 2595
Smältpunkt: 1083
Densitet: 8.94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545