Koppar ≥99.999%, stavar, Puratronic®, Ø 7 mm (0.3 in)
Leverantör: Thermo Fisher Scientific
11443.DO 11443.HT
11443.DOEA 8700 SEK
11443.DO 11443.HT
Koppar ≥99.999%, stavar, Puratronic®, Ø 7 mm (0.3 in)
Koppar
Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈3.44g/cm Odour: Odourless MDL No.: MFCD00010965
Formel: Cu
Kokpunkt: 2595 °C (1013 hPa)
Smältpunkt: 1083 °C
Densitet: 8,94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545