Koppar ≥99.999%, stavar, Puratronic®, Length: random, Ø 3.18 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
00761.14 00761.22
00761.14EA 3075 SEK
00761.14 00761.22
Koppar ≥99.999%, stavar, Puratronic®, Length: random, Ø 3.18 mm (0.125 in)
Koppar
Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈0.71g/cm Odour: Odourless MDL No.: MFCD00010965
Formel: Cu
Kokpunkt: 2595 °C (1013 hPa)
Smältpunkt: 1083 °C
Densitet: 8,94 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545