Lantan 99,9 REO basis, pieces
Leverantör: Thermo Fisher Scientific
Fara
00175.14 00175.22
00175.14EA 3775 SEK
00175.14 00175.22
Lantan 99,9 REO basis, pieces
Lantan
Purity: 99.9% CAS No.: 7439-91-0 Physical Form: Packaged under mineral oil Appearance: Silver-grey UN No.: 3208 MDL No.: MFCD00011066
Formel: La
MW: 138.91 g/mol
Kokpunkt: 3464
Smältpunkt: 920
Densitet: ∼6.2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011066
CAS nummer: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3 II