Jod, EMPROVE® ESSENTIAL USP, BP suitable for use as excipient, sublimeras, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
M-Clarity™ quality level = MQ100
1.04760.2500 1.04760.9050
1.04760.2500EA 36775 SEK
1.04760.2500 1.04760.9050
Jod, EMPROVE® ESSENTIAL USP, BP suitable for use as excipient, sublimeras, SAFC®
Jod
Formel: I₂
MW: 253.81 g/mol
Kokpunkt: 184.3
Smältpunkt: 114
Densitet: 4.9 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011355
CAS nummer: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 (6.1) III
Merck Index: 13,05036