Tellur ≥99.999%, pulver, Puratronic® -18/+60 mesh

Leverantör: Thermo Scientific
Fara

43960.18 43960.30
43960.18EA 3500 SEK
43960.18 43960.30
Tellur ≥99.999%, pulver, Puratronic® -18/+60 mesh
Tellurium

Purity: 99.999% CAS No.: 13494-80-9 Odour: Odourless UN No.: 3288 MDL No.: MFCD00134062

Formel: Te
MW: 127,6 g/mol
Kokpunkt: 990 °C (1013 hPa)
Smältpunkt: 450 °C
Densitet: 8,83 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134062
CAS nummer: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,09273

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR